Learn More
Grainger Drill and Tap Set 8 -32NC Thread 2 pcs.
Catalog No. 502752476
Encompass_Preferred
The Grainger Approved 8-32NC Drill and Tap Set is a high-speed steel tap and drill set for creating 8-32NC threaded holes in metal, plastic, and other materials. It includes a #29 drill bit and an 8-32NC tap with 3 flutes. The tap has a spiral point for easy starting and cutting threads. The drill and tap are made from high-speed steel for durability and have black oxide finish to resist corrosion. This is a complete set for drilling pilot holes and tapping internal threads sized for 8-32 machine screws.
Catalog No. 50-275-2476 Supplier Grainger Supplier No. 141H64
The Grainger Approved 8-32NC Drill and Tap Set is a high-speed steel tap and drill set for creating 8-32NC threaded holes in metal, plastic, and other materials. It includes a #29 drill bit and an 8-32NC tap with 3 flutes. The tap has a spiral point for easy starting and cutting threads. The drill and tap are made from high-speed steel for durability and have black oxide finish to resist corrosion. This is a complete set for drilling pilot holes and tapping internal threads sized for 8-32 machine screws.