Learn More
Stoelting™ Drill Bit
SDP
Catalog No. 10001090 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
0.60 mm
0.75 mm
1 mm
1.35 mm
1.75 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-001-090 1.75 mm
10-001-086 0.60 mm
10-001-087 0.75 mm
10-001-088 1 mm
10-001-089 1.35 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-001-090 Supplier Stoelting™ Supplier No. 514556
Add to Cart
Add to Cart
Stoelting™ Carbide Drill Bits cut cleaner and retain sharp edges longer.
  • Generally used in Stereotaxic Surgery and procedures
  • Suitable for use with the 58650 Flexible Shaft Drill, and the 51449 Micromotor Drill
  • Spherical heads, variety of sizes
  • Packages of 6 bits (all the same size)
Specifications
For Use With (Application) Neuroscience
Size 1.75 mm
Material Carbide