Learn More
Grainger Huot Stub Screw Machine Drill Index, 60 Compartments, #1 to #60, 1 ea
Catalog No. 502788140
Encompass_Preferred
This stub screw machine index is a specialized steel storage solution designed for organizing 60 individual drill bits within a wire size range of #1 to #60. Finished with a durable epoxy coating, this index provides clear, organized access to commonly used bits, making it a functional addition to any workshop or tool storage setup.
  • Organizes up to 60 stub length drill bits
  • Accommodates wire sizes from #1 to #60
  • Constructed from steel with an epoxy finish
  • Includes decimal equivalents and screw tap information
  • Measures 3-1/4 in. x 4-1/2 in. x 1 in.
Catalog No. 50-278-8140 Supplier Grainger Supplier No. 18EC46
This stub screw machine index is a specialized steel storage solution designed for organizing 60 individual drill bits within a wire size range of #1 to #60. Finished with a durable epoxy coating, this index provides clear, organized access to commonly used bits, making it a functional addition to any workshop or tool storage setup.
  • Organizes up to 60 stub length drill bits
  • Accommodates wire sizes from #1 to #60
  • Constructed from steel with an epoxy finish
  • Includes decimal equivalents and screw tap information
  • Measures 3-1/4 in. x 4-1/2 in. x 1 in.