Learn More
Grainger Drill Mill 1/4 Carbide Bright
Catalog No. 502766940
Encompass_Preferred
The Micro 100 Drill Mill Carbide is a high-performance bit for milling and drilling in metals like steel, cast iron, and aluminum. It features a TiAlN coating for longer tool life and heat resistance, and a 135-degree split point tip to reduce walking. Made of micrograin carbide, it is strong and durable. The bit is 3/8 inch in diameter, 2-1/2 inches long, with a 1/2 inch shank, suitable for high-speed machining. It can machine alloy steels, tool steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and composites. Made in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6940 Supplier Grainger Supplier No. 16T581
The Micro 100 Drill Mill Carbide is a high-performance bit for milling and drilling in metals like steel, cast iron, and aluminum. It features a TiAlN coating for longer tool life and heat resistance, and a 135-degree split point tip to reduce walking. Made of micrograin carbide, it is strong and durable. The bit is 3/8 inch in diameter, 2-1/2 inches long, with a 1/2 inch shank, suitable for high-speed machining. It can machine alloy steels, tool steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and composites. Made in the USA by Micro 100 Tool Corp.