Learn More
Grainger Drill Mill 1/8 Carbide AlTiN
Catalog No. 502766938
Encompass_Preferred
The Grainger 16T574 is a Dayton 1/3 HP Direct Drive Blower Motor that runs on 115V AC power at 1725 RPM. It has a 48 frame size, is single phase, and is totally enclosed with fan cooled construction. The motor has class B insulation and a continuous duty rating. It can provide airflow up to 850 CFM. The blower outlet diameter is 6". This electric blower motor is suitable for air moving applications like cooling electrical enclosures or ventilating equipment.
Catalog No. 50-276-6938 Supplier Grainger Supplier No. 16T574
The Grainger 16T574 is a Dayton 1/3 HP Direct Drive Blower Motor that runs on 115V AC power at 1725 RPM. It has a 48 frame size, is single phase, and is totally enclosed with fan cooled construction. The motor has class B insulation and a continuous duty rating. It can provide airflow up to 850 CFM. The blower outlet diameter is 6". This electric blower motor is suitable for air moving applications like cooling electrical enclosures or ventilating equipment.