Learn More
Grainger Drill Mill 3/16 Carbide AlTiN
Catalog No. 502766811
Encompass_Preferred
The Grainger 16T576 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with its compact size and enclosed hydraulic system. The jack's lift range makes it suitable for use with light and medium duty automotive lifting applications. Its hydraulic power system enables smooth lifting up to 16 tons with minimal effort. The jack meets ASME PALD standards.
Catalog No. 50-276-6811 Supplier Grainger Supplier No. 16T576
The Grainger 16T576 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a minimum height of 10-3/4 inches and a maximum height of 16-1/4 inches. The jack has a welded base for stability with rubber pads to prevent slipping. It is designed for portability with its compact size and enclosed hydraulic system. The jack's lift range makes it suitable for use with light and medium duty automotive lifting applications. Its hydraulic power system enables smooth lifting up to 16 tons with minimal effort. The jack meets ASME PALD standards.