Learn More
Grainger Drill Mill 3/16 Carbide AlTiN
Catalog No. 502766631
Encompass_Preferred
The Grainger Approved 3/16" Carbide AlTiN Drill Mill is a high-performance cutting tool designed for milling and drilling operations. It features a 135 degree split point tip to reduce thrust and wandering. The AlTiN coating provides heat and wear resistance. It has a 3/16" diameter and is available in lengths ranging from 1" to 4" in 1/4" increments. The shank diameter is 1/4". This USA-made solid carbide tool is rigid and precise for excellent hole quality in steel, cast iron, aluminum and other non-ferrous metals.
Catalog No. 50-276-6631 Supplier Grainger Supplier No. 16T578
The Grainger Approved 3/16" Carbide AlTiN Drill Mill is a high-performance cutting tool designed for milling and drilling operations. It features a 135 degree split point tip to reduce thrust and wandering. The AlTiN coating provides heat and wear resistance. It has a 3/16" diameter and is available in lengths ranging from 1" to 4" in 1/4" increments. The shank diameter is 1/4". This USA-made solid carbide tool is rigid and precise for excellent hole quality in steel, cast iron, aluminum and other non-ferrous metals.