Learn More
Grainger Drill Mill 3/16 Carbide Bright
Catalog No. 502764934
Encompass_Preferred
The WIDIA HANITA 2NHF7 is a carbide drill mill with a diameter of 0.3937 inches (10mm) and an overall length of 2.559 inches (65mm). It has a TiAlN coating and can be used for a variety of materials including steel, cast iron, stainless steel, hardened steel, titanium alloys, nickel alloys, and non-ferrous metals. The drill mill features a 135 degree split point tip geometry for reduced thrust and chatter-free cutting. It has internal coolant capability and is balanced to 25,000 RPM. The 2NHF7 is manufactured in Israel by WIDIA HANITA Cutting Tools Ltd.
Catalog No. 50-276-4934 Supplier Grainger Supplier No. 16T577
The WIDIA HANITA 2NHF7 is a carbide drill mill with a diameter of 0.3937 inches (10mm) and an overall length of 2.559 inches (65mm). It has a TiAlN coating and can be used for a variety of materials including steel, cast iron, stainless steel, hardened steel, titanium alloys, nickel alloys, and non-ferrous metals. The drill mill features a 135 degree split point tip geometry for reduced thrust and chatter-free cutting. It has internal coolant capability and is balanced to 25,000 RPM. The 2NHF7 is manufactured in Israel by WIDIA HANITA Cutting Tools Ltd.