Learn More
Grainger Drill Mill 3/4 Carbide Bright
Catalog No. 502766399
Encompass_Preferred
The Grainger 16T606 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.
Catalog No. 50-276-6399 Supplier Grainger Supplier No. 16T606
The Grainger 16T606 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of power with 1.8 nominal amps at full load. This electric motor is designed for general purpose applications.