Learn More
Grainger Drill Mill 3/4 Carbide Bright
Catalog No. 502766413
Encompass_Preferred
The Micro 100 16T604 is a carbide drill mill with a 1/4" shank diameter and a 1/2" cutting diameter. It has a TiAlN coating and can be run at speeds up to 325 SFM. The drill mill has a 135 degree split point tip angle and is made of micrograin carbide for strength and heat resistance. It can drill, chamfer, face mill and slot in steel, cast iron, aluminum and other non-ferrous metals. The 16T604 is manufactured in the USA by Micro 100 Tool Corp.
Catalog No. 50-276-6413 Supplier Grainger Supplier No. 16T604
The Micro 100 16T604 is a carbide drill mill with a 1/4" shank diameter and a 1/2" cutting diameter. It has a TiAlN coating and can be run at speeds up to 325 SFM. The drill mill has a 135 degree split point tip angle and is made of micrograin carbide for strength and heat resistance. It can drill, chamfer, face mill and slot in steel, cast iron, aluminum and other non-ferrous metals. The 16T604 is manufactured in the USA by Micro 100 Tool Corp.