Learn More
Grainger Drill Mill 3/8 Carbide AlTiN
Catalog No. 502766397
Encompass_Preferred
The Grainger Approved 3/8" Carbide Drill Mill is a high-speed steel cutting tool designed for milling and drilling operations. It features a 135 degree split point tip for faster penetration and reduced walking. The drill has a 3/8" diameter, is 6" overall length, and has a 1/2" shank diameter. It is coated in titanium nitride for increased hardness and lubricity. The drill mill is manufactured in the USA and ideal for use on steel, cast iron, aluminum and other non-ferrous metals. It provides good chip clearance while drilling and milling.
Catalog No. 50-276-6397 Supplier Grainger Supplier No. 16T590
The Grainger Approved 3/8" Carbide Drill Mill is a high-speed steel cutting tool designed for milling and drilling operations. It features a 135 degree split point tip for faster penetration and reduced walking. The drill has a 3/8" diameter, is 6" overall length, and has a 1/2" shank diameter. It is coated in titanium nitride for increased hardness and lubricity. The drill mill is manufactured in the USA and ideal for use on steel, cast iron, aluminum and other non-ferrous metals. It provides good chip clearance while drilling and milling.