Learn More
Grainger Drill Mill 3/8 Carbide AlTiN
Catalog No. 502764949
Encompass_Preferred
The Micro 100 Drill Mill Carbide is a high-performance bit for milling and drilling in metals like steel, cast iron, and aluminum. It features a TiAlN coating for durability and heat resistance, and a 135-degree split point tip to minimize walking. Made of micrograin carbide, it measures 3/8 inch in diameter, 2-1/2 inches long, with a 1/2 inch shank. Suitable for high-speed machining, it handles alloy steels, tool steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and composites. Made in the USA by Micro 100 Tool Company.
Catalog No. 50-276-4949 Supplier Grainger Supplier No. 16T598
The Micro 100 Drill Mill Carbide is a high-performance bit for milling and drilling in metals like steel, cast iron, and aluminum. It features a TiAlN coating for durability and heat resistance, and a 135-degree split point tip to minimize walking. Made of micrograin carbide, it measures 3/8 inch in diameter, 2-1/2 inches long, with a 1/2 inch shank. Suitable for high-speed machining, it handles alloy steels, tool steels, cast iron, stainless steel, aluminum, copper, brass, bronze, plastics, and composites. Made in the USA by Micro 100 Tool Company.