Learn More
Grainger Drill Mill 5/8 Carbide Bright
Catalog No. 502765909
Encompass_Preferred
The Drill Mill Carbide 16T602 is a carbide end mill with a diameter of 1/2 inch and an overall length of 2 inches. It has a TiAlN coating and 4 flutes. The end mill is made of solid carbide and can machine steels, stainless steels, cast iron, hardened steels, titanium alloys, nickel alloys, and other non-ferrous metals. It has a center cutting design for plunging. Recommended speed is 500-4000 RPM with feed rates from 0.001-0.060 IPR based on material. It has an ATI2 coating that resists heat and wear. The end mill has a tolerance of +/- 0.0005 inches and a corner radius of 1/64 inch.
Catalog No. 50-276-5909 Supplier Grainger Supplier No. 16T602
The Drill Mill Carbide 16T602 is a carbide end mill with a diameter of 1/2 inch and an overall length of 2 inches. It has a TiAlN coating and 4 flutes. The end mill is made of solid carbide and can machine steels, stainless steels, cast iron, hardened steels, titanium alloys, nickel alloys, and other non-ferrous metals. It has a center cutting design for plunging. Recommended speed is 500-4000 RPM with feed rates from 0.001-0.060 IPR based on material. It has an ATI2 coating that resists heat and wear. The end mill has a tolerance of +/- 0.0005 inches and a corner radius of 1/64 inch.