Learn More
World Precision Instrument Drill with Drill Bits for Premium Motor Stereotaxic
SDP
Catalog No. 502539079
Encompass
New portable micromotor with pocket/pouch-size control unit has built-in lithium-ion rechargeable battery for the ultimate in portability. The control unit features a digital RPM display at the top that makes it easy to monitor speed as you work. The dial on the front adjusts speed from 0-30,000 RPM.
Catalog No. 50-253-9079 Supplier World Precision Instrument Supplier No. 505379
New portable micromotor with pocket/pouch-size control unit has built-in lithium-ion rechargeable battery for the ultimate in portability. The control unit features a digital RPM display at the top that makes it easy to monitor speed as you work. The dial on the front adjusts speed from 0-30,000 RPM.