Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Drilled Balls, Set of 14
SCI_ED

Catalog No. S139417 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139417 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139417 Supplier Supertek™ Supplier No. PH10027SP
Add to Cart
Add to Cart

Ideal for collision and pendulum applications. Includes two balls each of aluminum, brass, copper, iron, lead, zinc, and wood.

  • Handy 1 in. pre-drilled balls.
  • 14 balls of 7 materials.
  • 2 aluminum balls
  • 2 brass balls
  • 2 copper balls
  • 2 iron balls
  • 2 lead balls
  • li>2 zinc balls
  • 2 wood balls

Specifications

Grade 5+
Format Set
Languages English
Subject Physical Science - Force and Mass
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.