Learn More
Grainger Drilling Jig 80/20 10 Series
Supplier:  Grainger 16U338
Encompass_Preferred
The 80/20 Drilling Jig 10 Series is a versatile drilling fixture that allows for quick and accurate hole placement. It is constructed from aluminum extrusion and features an anodized coating for durability. The jig has a 10" x 10" grid pattern with 1" hole spacing, providing up to 100 possible drill locations. It comes with an adjustable fence with scales for precision setups and can handle material up to 2" thick. The jig works with drill presses, mills and lathes to ensure perpendicularity when drilling holes. Its rigid design eliminates deflection during operation. The 80/20 Drilling Jig 10 Series enables efficient, precise hole drilling for various applications.
Catalog No. 50-276-6835