Learn More
TARGETMOL CHEMICALS INC DRINABANT 5MG
SDP
Catalog No. 502580995
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Drinabant (AVE-1625) is an orally active CB1 receptor antagonist. Drinabant inhibits the agonist-stimulated calcium signal with IC50 values of 25 nM and 10 nM for the hCB1-R and rCB1-R respectively and is ineffective for the hCB2-R. purity: 99%
Catalog No. 50-258-0995 Supplier TARGETMOL CHEMICALS INC Supplier No. T218615MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Drinabant (AVE-1625) is an orally active CB1 receptor antagonist. Drinabant inhibits the agonist-stimulated calcium signal with IC50 values of 25 nM and 10 nM for the hCB1-R and rCB1-R respectively and is ineffective for the hCB2-R. purity: 99%