Learn More
MSC Envu Insecticide Dust, Desiccant, Pyrethroid, Broad Spectrum
Catalog No. 502838491
Encompass
This desiccant dust controls pests in commercial and industrial areas. Effective against ants, bedbugs, wasps, cockroaches, silverfish, and fleas, it offers up to 6 months of residual control. Its active ingredients, pyrethrum and amorphous silica gel, work by eroding insect cuticles, causing dehydration.
  • Excellent flushing action to pinpoint infestations.
  • Provides fast pest knockdown.
  • Offers long-lasting residual effect (up to six months).
  • Achieves superior control on porous surfaces.
  • Ensures deep, uniform penetration of voids.
  • Delivers broad-spectrum pest control.
  • Ready to use; no mixing required.
  • Lightweight and easy to carry.
  • Compatible with professional dust application equipment.
  • Ideal for carpenter ant inspections; flushes ants and pinpoints nest locations.
Catalog No. 50-283-8491 Supplier MSC Supplier No. 74336488
This desiccant dust controls pests in commercial and industrial areas. Effective against ants, bedbugs, wasps, cockroaches, silverfish, and fleas, it offers up to 6 months of residual control. Its active ingredients, pyrethrum and amorphous silica gel, work by eroding insect cuticles, causing dehydration.
  • Excellent flushing action to pinpoint infestations.
  • Provides fast pest knockdown.
  • Offers long-lasting residual effect (up to six months).
  • Achieves superior control on porous surfaces.
  • Ensures deep, uniform penetration of voids.
  • Delivers broad-spectrum pest control.
  • Ready to use; no mixing required.
  • Lightweight and easy to carry.
  • Compatible with professional dust application equipment.
  • Ideal for carpenter ant inspections; flushes ants and pinpoints nest locations.