Learn More
Grainger Drip Emitter Conversion Kit
Catalog No. 502751769
Encompass_Preferred
The Rain Bird Drip Emitter Conversion Kit converts existing Rain Bird spray heads or rotors to drip irrigation. The kit includes a 1/2" female threaded adapter, a pressure compensating emitter, and tubing. It provides 2 gallons per hour flow at 25 PSI. The kit allows converting spray heads to drip irrigation without having to replace the entire system. It is useful for watering individual plants or for targeted irrigation. The pressure compensating emitter helps provide consistent flow even when pressure fluctuates. The kit works with most Rain Bird spray heads and rotors. It provides an easy way to convert parts of an existing sprinkler system to low volume drip irrigation.
Catalog No. 50-275-1769 Supplier Grainger Supplier No. 14U326
The Rain Bird Drip Emitter Conversion Kit converts existing Rain Bird spray heads or rotors to drip irrigation. The kit includes a 1/2" female threaded adapter, a pressure compensating emitter, and tubing. It provides 2 gallons per hour flow at 25 PSI. The kit allows converting spray heads to drip irrigation without having to replace the entire system. It is useful for watering individual plants or for targeted irrigation. The pressure compensating emitter helps provide consistent flow even when pressure fluctuates. The kit works with most Rain Bird spray heads and rotors. It provides an easy way to convert parts of an existing sprinkler system to low volume drip irrigation.