Learn More
Waters Corp Waters Drip Tray, Fraction Collector III, 8.0 mm OD Drain
Catalog No. 50792696 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Drip Tray is an optional component for the Waters Fraction Collector III, designed for stand-alone installations where the instrument’s arm may drain fluid past the rack. It effectively collects fluids and can also function as a collection reservoir when an optional drain tube plug is utilized.
  • Easily installs by sliding under the front of the unit and aligning with rubber feet.
  • Connects to 8.0 mm OD drain tubing for efficient fluid discharge.
  • Functions as a collection reservoir with an optional drain tube plug.
Catalog No. 50-792-696 Supplier Waters Corp Supplier No. 700002164
The Waters Drip Tray is an optional component for the Waters Fraction Collector III, designed for stand-alone installations where the instrument’s arm may drain fluid past the rack. It effectively collects fluids and can also function as a collection reservoir when an optional drain tube plug is utilized.
  • Easily installs by sliding under the front of the unit and aligning with rubber feet.
  • Connects to 8.0 mm OD drain tubing for efficient fluid discharge.
  • Functions as a collection reservoir with an optional drain tube plug.