Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ Drip Tray, XPR/XSR Analytical Balance

Catalog No. 13940080 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Drip Tray, XPR/XSR Analytical Balance
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-940-080 Drip Tray, XPR/XSR Analytical Balance
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-940-080 Supplier METTLER TOLEDO™ Supplier No. 30460856
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Drip pan for better visibility of light colored powders and stains for XPR/XSR analytical balances

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) XPR/XSR Analytical Balance
Color Gray
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.