Learn More
Hach Company Tube, Drip
Supplier:  Hach Company 2265100
Encompass
Borosilicate glass drip tube with 7 mm outside diameter, 45° bevel tip. Body is 17 mm outside diameter and 145 mm in length with a 20 to 400 screw-thread finish. Use on Hach Distillation Apparatus.
Catalog No. 50-199-5672