Learn More
Medchemexpress LLC Driselase from Basidiomycetes sp. | 85186-71-6 | MFCD00130978 | >10.0% | 100MG
SDP
Catalog No. 5000239607
Encompass_Preferred
Driselase from Basidiomycetes sp. is a commercial enzyme preparation composed of multiple fungal carbohydrolase activities used to digest plant cell walls and facilitate protoplast formation. Supplied as a white to yellow solid powder, it is soluble in water and formulated for enzymatic hydrolysis of cellulose, hemicellulose, and pectin components in research applications.
Catalog No. 50-002-39607 Supplier Medchemexpress LLC Supplier No. HY126224100MG
Driselase from Basidiomycetes sp. is a commercial enzyme preparation composed of multiple fungal carbohydrolase activities used to digest plant cell walls and facilitate protoplast formation. Supplied as a white to yellow solid powder, it is soluble in water and formulated for enzymatic hydrolysis of cellulose, hemicellulose, and pectin components in research applications.