Learn More
Kuhner Shaker Inc DRIVE CONTROLLER FOR SB50-X S
Catalog No. 502569958
Encompass
Shaking machine OrbShakeTemperature range with cooling:Low: Depends on cooling liquid and cultureHigh: 50C50mm orbital diameter w/ speeds up to 150rpmControlled acceleration rate includedActive and Positional braking: included and adjustableTimer included and adjustableDC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H1940 x W1013 x D991mm; Weight: 340kgFIVE YEAR WARRANTY HARDWARE
Catalog No. 50-256-9958 Supplier Kuhner Shaker Inc Supplier No. SMX7500
Add to Cart
Add to Cart
Request a Quote
Shaking machine OrbShakeTemperature range with cooling:Low: Depends on cooling liquid and cultureHigh: 50C50mm orbital diameter w/ speeds up to 150rpmControlled acceleration rate includedActive and Positional braking: included and adjustableTimer included and adjustableDC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorDimensions: H1940 x W1013 x D991mm; Weight: 340kgFIVE YEAR WARRANTY HARDWARE