Learn More
AVANTOR MASTERFLEX (VWR LLC) DRIVE MFLEX BRUSHLESS 115/230V
Catalog No. 502742129
Encompass
Masterflex I/P Brushless Process Drive, 33 to 650 rpm; 115/230 VAC, Avantor
Catalog No. 50-274-2129 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7741010
Masterflex I/P Brushless Process Drive, 33 to 650 rpm; 115/230 VAC, Avantor