Learn More
AVANTOR MASTERFLEX (VWR LLC) DRIVE MFLEX I/P 650R SS E/IP
Catalog No. 502742128
Encompass
Masterflex I/P MasterSense™ Drive with EtherNet/IP™ and MasterflexLive, 0.1 to 650 rpm; 115/230 VAC, Avantor
Catalog No. 50-274-2128 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7742100
Add to Cart
Add to Cart
Request a Quote
Masterflex I/P MasterSense™ Drive with EtherNet/IP™ and MasterflexLive, 0.1 to 650 rpm; 115/230 VAC, Avantor