Learn More
AVANTOR MASTERFLEX (VWR LLC) DRIVE MFLEX I/P 650R SS PROF
Catalog No. 502742127
Encompass
Masterflex I/P MasterSense™ Drive with Profibus and MasterflexLive, 0.1 to 650 rpm; 115/230 VAC, Avantor
Catalog No. 50-274-2127 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7742102
Add to Cart
Add to Cart
Request a Quote
Masterflex I/P MasterSense™ Drive with Profibus and MasterflexLive, 0.1 to 650 rpm; 115/230 VAC, Avantor