Learn More
AVANTOR MASTERFLEX (VWR LLC) DRIVE MFLEX L/S 600R SS E/IP
Catalog No. 502742792
Encompass
Masterflex L/S MasterSense™ Drive with EtherNet/IP and MasterflexLive, Stainless Steel Housing, 0.1 to 600 rpm; 115/230 VAC, Avantor
Catalog No. 50-274-2792 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0757610
Add to Cart
Add to Cart
Request a Quote
Masterflex L/S MasterSense™ Drive with EtherNet/IP and MasterflexLive, Stainless Steel Housing, 0.1 to 600 rpm; 115/230 VAC, Avantor