Learn More
AVANTOR MASTERFLEX (VWR LLC) DRIVE MFLEX L/S 600RPM 115/230
Catalog No. 502742793
Encompass
Masterflex L/S Digital Process Drive, Stainless Steel Housing, 0.1 to 600 rpm; 90 to 260 VAC, Avantor
Catalog No. 50-274-2793 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0757530
Add to Cart
Add to Cart
Request a Quote
Masterflex L/S Digital Process Drive, Stainless Steel Housing, 0.1 to 600 rpm; 90 to 260 VAC, Avantor