Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Variable-Speed, Compact, Drive, 35 to 200 rpm; 12 VDC
Catalog No. NC0161753
Encompass
These compact drives are a perfect when space is limited, as they fit just about anywhere. The variable-speed design makes them ideal for when you need to pump fluid at a higher rate. Drives accept one Easy-Load or standard pump head. Use drives with L/S Precision tubing¿accepts sizes L/S 13, L/S 14, L/S 16, L/S 25, and L/S 17. Only silicone and C-Flex tubing formulations are recommended for use with L/S 25 and L/S 17 sizes.
Catalog No. NC0161753 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720012
These compact drives are a perfect when space is limited, as they fit just about anywhere. The variable-speed design makes them ideal for when you need to pump fluid at a higher rate. Drives accept one Easy-Load or standard pump head. Use drives with L/S Precision tubing¿accepts sizes L/S 13, L/S 14, L/S 16, L/S 25, and L/S 17. Only silicone and C-Flex tubing formulations are recommended for use with L/S 25 and L/S 17 sizes.