Learn More
Uline Drum Overpack Standard
Supplier:  Uline H10740
Encompass
Standard 22 gallon by 19 inch drum overpack containment system for safely enclosing damaged or leaking drums to prevent further spillage and exposure.
Catalog No. NC2311860