Learn More
Electron Microscopy Sciences DrainRack™ Junior Drain Rack, Blue
SDP
Catalog No. 501899570
Encompass_Preferred
Made from high impact polystyrene This model is similar to the DrainRack above but can hold up to 100 microscope slides in 50 individual numbered slots. The rack is autoclavable.
Color: Blue
Catalog No. 50-189-9570 Supplier Electron Microscopy Sciences Supplier No. 71445B
Made from high impact polystyrene This model is similar to the DrainRack above but can hold up to 100 microscope slides in 50 individual numbered slots. The rack is autoclavable.
Color: Blue