Learn More
Electron Microscopy Sciences DrainRack™ Junior Drain Rack, Yellow
SDP
Catalog No. 501899574
Encompass_Preferred
Made from high impact polystyrene This model is similar to the DrainRack above but can hold up to 100 microscope slides in 50 individual numbered slots. The rack is autoclavable.
Color: Yellow
Catalog No. 50-189-9574 Supplier Electron Microscopy Sciences Supplier No. 71445Y
Made from high impact polystyrene This model is similar to the DrainRack above but can hold up to 100 microscope slides in 50 individual numbered slots. The rack is autoclavable.
Color: Yellow