Learn More
Electron Microscopy Sciences DrainRack™ Microscope Slide Drain Rack, Green
SDP
Catalog No. 501899576
Encompass_Preferred
Made from high impact polystyrene. This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individual numbered slots. All sides are stored upright for easy insertion and removal. A unique feature with the DrainRack is the ability to read bar codes without having to remove the slides from the rack.
For space saving purposes, you can double the amount of slides simply by storing 2 slides per slot, giving you a capacity of 200 slides per tray instead of 100. The racks are not autoclavable. They measure: 75 mm x 231 mm x 25 mm H (3"x 9"x1" H)
Color: Green
Catalog No. 50-189-9576 Supplier Electron Microscopy Sciences Supplier No. 71446G
Made from high impact polystyrene. This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individual numbered slots. All sides are stored upright for easy insertion and removal. A unique feature with the DrainRack is the ability to read bar codes without having to remove the slides from the rack.
For space saving purposes, you can double the amount of slides simply by storing 2 slides per slot, giving you a capacity of 200 slides per tray instead of 100. The racks are not autoclavable. They measure: 75 mm x 231 mm x 25 mm H (3"x 9"x1" H)
Color: Green