Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRO Mixture (EPA/Wisconsin), 2000μg/mL in dichloromethane, Ultra Scientific

Catalog No. USUST2001
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-UST-200-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-UST-200-1 Supplier Agilent Technologies Supplier No. UST2001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material DRO Mixture (EPA/Wisconsin)
CAS Max % 98.49, 0.15, 0.15, 0.15, 0.15, 0.15
CAS 124-18-5, 112-40-3, 629-59-4, 544-76-3, 593-45-3
Analyte or Component Names Methylene chloride (dichloromethane),n-decane,n-dodecane
Element Methylene Chloride, n-decane, n-dodecane
Solvent or Matrix Dichloromethane
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.