Learn More
Selleck Chemical LLC Drofenine Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S566525MG
Encompass_Preferred
Drofenine hydrochloride is a potent competitive inhibitor of BChE It is an anticholinergic that acts both at the level of the nerves and the muscle fibers
Catalog No. 50-000-03313