Learn More
Medchemexpress LLC Drofenine hydrochloride | 548-66-3 | 98.1% | 1 ML
SDP
Catalog No. 5000412867
Encompass_Preferred
Drofenine hydrochloride is a potent competitive cholinesterase inhibitor. It acts as a BChE inhibitor with a Ki value of 3 μM and is intended for research use only.
  • Potent competitive cholinesterase inhibitor
  • BChE inhibitor
  • Ki value of 3 μM
  • Suitable for research use
Catalog No. 50-004-12867 Supplier Medchemexpress LLC Supplier No. HYB123910MM/1ML
Drofenine hydrochloride is a potent competitive cholinesterase inhibitor. It acts as a BChE inhibitor with a Ki value of 3 μM and is intended for research use only.
  • Potent competitive cholinesterase inhibitor
  • BChE inhibitor
  • Ki value of 3 μM
  • Suitable for research use