Learn More
TARGETMOL CHEMICALS INC DROLOXIFENE 5MG
SDP
Catalog No. 502581575
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Droloxifene (3-Hydroxytamoxifen) a tamoxifen derivative is an oral active and selective estrogen receptor modulator.Droloxifene induced p53 expression and apoptosis in McF-7 cells.Fluroxifene has anti-estrogen and anti-implantation effects.Fluroxifene prevented bone loss in ovariectomized rats. purity: 99%
Catalog No. 50-258-1575 Supplier TARGETMOL CHEMICALS INC Supplier No. T110985MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Droloxifene (3-Hydroxytamoxifen) a tamoxifen derivative is an oral active and selective estrogen receptor modulator.Droloxifene induced p53 expression and apoptosis in McF-7 cells.Fluroxifene has anti-estrogen and anti-implantation effects.Fluroxifene prevented bone loss in ovariectomized rats. purity: 99%