Learn More
Cayman Chemical Dronedarone (hydrochloride), Purity: ≥98%, CAS: 141625-93-6, 100 mg
SDP
Catalog No. 502894883
Encompass_Preferred
An antiarrhythmic agent; inhibits muscarinic acetylcholine receptor-dependent potassium currents induced by carbachol or intracellular loading of GTPγS in single cells isolated from guinea pig atria (IC50s = 0.1 and 1 µM, respectively); reduces sinus frequency and prolongs APD in isolated rabbit ventricular myocardium at 50 and 100 mg/kg; increases atrial APD and the AERP in a dog model of atrial fibrillation
Catalog No. 50-289-4883 Supplier Cayman Chemical Supplier No. 9000543100 MG
An antiarrhythmic agent; inhibits muscarinic acetylcholine receptor-dependent potassium currents induced by carbachol or intracellular loading of GTPγS in single cells isolated from guinea pig atria (IC50s = 0.1 and 1 µM, respectively); reduces sinus frequency and prolongs APD in isolated rabbit ventricular myocardium at 50 and 100 mg/kg; increases atrial APD and the AERP in a dog model of atrial fibrillation