Learn More
Cayman Chemical DrondaronhydrochlorIde 50mg
SDP
Catalog No. 502892508
Encompass_Preferred
An antiarrhythmic agent; inhibits muscarinic acetylcholine receptor-dependent potassium currents induced by carbachol or intracellular loading of GTPγS in single cells isolated from guinea pig atria (IC50s = 0.1 and 1 µM, respectively); reduces sinus frequency and prolongs APD in isolated rabbit ventricular myocardium at 50 and 100 mg/kg; increases atrial APD and the AERP in a dog model of atrial fibrillation
Catalog No. 50-289-2508 Supplier Cayman Chemical Supplier No. 900054350 MG
An antiarrhythmic agent; inhibits muscarinic acetylcholine receptor-dependent potassium currents induced by carbachol or intracellular loading of GTPγS in single cells isolated from guinea pig atria (IC50s = 0.1 and 1 µM, respectively); reduces sinus frequency and prolongs APD in isolated rabbit ventricular myocardium at 50 and 100 mg/kg; increases atrial APD and the AERP in a dog model of atrial fibrillation