Learn More
Apexbio Technology LLC Dronedarone 141626-36-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A33745.1
Encompass_Preferred
Dronedarone (CAS 141626-36-0) is a benzofuran derivative structurally related to amiodarone designed to manage cardiac arrhythmias It exerts its pharmacological effects primarily through multichannel blockade inhibiting sodium potassium and calcium ion currents and demonstrating anti-adrenergic properties By prolonging action potential duration and refractory periods dronedarone contributes to the stabilization and restoration of regular cardiac rhythm In biomedical research contexts dronedarone serves as a valuable tool for exploring arrhythmogenesis mechanisms and developing therapeutic approaches for atrial fibrillation and atrial flutter
Catalog No. 50-000-62831