Learn More
Medchemexpress LLC Dronedarone Hydrochloride | 141625-93-6 | 99.99% | C31H45ClN2O5S | 100 MG
SDP
Catalog No. 5000434341
Encompass_Preferred
Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+, and Ca2+ currents. It serves as a multichannel blocker for atrial fibrillation, making it suitable for various research applications.
  • Inhibits Na+, K+, and Ca2+ currents.
  • Potently blocks acetylcholine-activated K+ current.
  • Blocks IKr and ICa-L with specific IC50 values.
  • Exhibits state-dependent inhibition of fast Na+ channel current.
  • Significantly reduces incidence of ventricular fibrillation in vivo.
  • Inhibits carotid artery thrombus formation in vivo.
Catalog No. 50-004-34341 Supplier Medchemexpress LLC Supplier No. HY75839100MG
Dronedarone Hydrochloride is a non-iodinated amiodarone derivative that inhibits Na+, K+, and Ca2+ currents. It serves as a multichannel blocker for atrial fibrillation, making it suitable for various research applications.
  • Inhibits Na+, K+, and Ca2+ currents.
  • Potently blocks acetylcholine-activated K+ current.
  • Blocks IKr and ICa-L with specific IC50 values.
  • Exhibits state-dependent inhibition of fast Na+ channel current.
  • Significantly reduces incidence of ventricular fibrillation in vivo.
  • Inhibits carotid artery thrombus formation in vivo.