Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Dronedarone hydrochloride, 98%

Catalog No. AAH3612403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36124-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH36124-03 Supplier Thermo Scientific™ Supplier No. H3612403
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 141625-93-6 | C31H45ClN2O5S | 593.22 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141625-93-6
Molecular Formula C31H45ClN2O5S
Molecular Weight (g/mol) 593.22
MDL Number MFCD00914940
InChI Key DWKVCQXJYURSIQ-UHFFFAOYSA-N
IUPAC Name hydrogen N-(2-butyl-3-{4-[3-(dibutylamino)propoxy]benzoyl}-1-benzofuran-5-yl)methanesulfonamide chloride
SMILES [H+].[Cl-].CCCCN(CCCC)CCCOC1=CC=C(C=C1)C(=O)C1=C(CCCC)OC2=CC=C(NS(C)(=O)=O)C=C12

Specifications

Melting Point 140-144°
Quantity 1 g
Formula Weight 593.22
Percent Purity 98%
Physical Form Solid
ShelfLife 5 Years
EINECSNumber 630-355-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.