Learn More
Med Vet International Drontal Plus, 22.7mg, 40 Taste Tabs
Catalog No. 502828378
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Drontal Plus (praziquantel/pyrantel pamoate/febantel) Taste Tabs broad spectrum chewable anthelmintic tablets are indicated for removal of Tapeworms (Dipylidium caninum, Taenia pisiformis, Echinococcus granulosus, and removal and control of Echinococcus multilocularis) and for removal of hookworms (Ancylostoma caninum, Uncinaria stenocephala), ascarids (Toxocara canis, Toxascaris leonina), and whipworms (Trichuris vulpis) in dogs. Most dogs find Drontal Plus Taste Tabs palatable. The tablets may be offered to the dog by hand. Alternatively, tablets may be given directly by mouth or offered in a small amount of food. Not for use in pregnant dogs or in puppies younger than 3 weeks or weighing less than 2 lbs. Each Drontal Plus Taste Tab for Puppies and Small Dogs contains 22.7 mg praziquantel, 22.7 mg pyrantel base as pyrantel pamoate and 113.4 mg febantel.
Catalog No. 50-282-8378 Supplier Med Vet International Supplier No. RXDRONK9SCHEW
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Drontal Plus (praziquantel/pyrantel pamoate/febantel) Taste Tabs broad spectrum chewable anthelmintic tablets are indicated for removal of Tapeworms (Dipylidium caninum, Taenia pisiformis, Echinococcus granulosus, and removal and control of Echinococcus multilocularis) and for removal of hookworms (Ancylostoma caninum, Uncinaria stenocephala), ascarids (Toxocara canis, Toxascaris leonina), and whipworms (Trichuris vulpis) in dogs. Most dogs find Drontal Plus Taste Tabs palatable. The tablets may be offered to the dog by hand. Alternatively, tablets may be given directly by mouth or offered in a small amount of food. Not for use in pregnant dogs or in puppies younger than 3 weeks or weighing less than 2 lbs. Each Drontal Plus Taste Tab for Puppies and Small Dogs contains 22.7 mg praziquantel, 22.7 mg pyrantel base as pyrantel pamoate and 113.4 mg febantel.