Learn More
MSC Trynex Tailgate Spreader, Tow Behind, Vehicle Battery-Powered, 14 cu ft Capacity
Catalog No. 502836378
Encompass
This vehicle battery-powered tailgate spreader is designed for dispensing de-icing pellets and granules onto walkways, driveways, and other paved areas. It offers a 14 cu ft capacity and is compatible with tractors.
  • Tow behind spreader type
  • 12V voltage
  • 1120 lb weight capacity
  • 14 cu ft capacity
  • Vehicle battery-powered
  • Compatible with tractor
  • For dispensing de-icing pellets and granules
  • Suitable for walkways, driveways, and other paved areas
Catalog No. 50-283-6378 Supplier MSC Supplier No. 57467060
Add to Cart
Add to Cart
Request a Quote
This vehicle battery-powered tailgate spreader is designed for dispensing de-icing pellets and granules onto walkways, driveways, and other paved areas. It offers a 14 cu ft capacity and is compatible with tractors.
  • Tow behind spreader type
  • 12V voltage
  • 1120 lb weight capacity
  • 14 cu ft capacity
  • Vehicle battery-powered
  • Compatible with tractor
  • For dispensing de-icing pellets and granules
  • Suitable for walkways, driveways, and other paved areas