Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Droperidol

Catalog No. AC460840010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460840010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC460840010 Supplier Thermo Scientific Chemicals Supplier No. 460840010

missing translation for 'validMainframeMsg'

CAS: 548-73-2 | C22H22FN3O2 | 379.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-73-2
Molecular Formula C22H22FN3O2
Molecular Weight (g/mol) 379.43
InChI Key RMEDXOLNCUSCGS-UHFFFAOYSA-N
Synonym droperidol, droleptan, inapsine, dehydrobenzperidol, properidol, sintodril, sintosian, inapsin, vetkalm, dridol
PubChem CID 3168
ChEBI CHEBI:4717
IUPAC Name 3-[1-[4-(4-fluorophenyl)-4-oxobutyl]-3,6-dihydro-2H-pyridin-4-yl]-1H-benzimidazol-2-one
SMILES C1CN(CC=C1N2C3=CC=CC=C3NC2=O)CCCC(=O)C4=CC=C(C=C4)F

Specifications

Melting Point 148.0°C to 149.0°C
Loss on Drying 5% max. (70°C, 4 h) (vaccuum)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in chlorofom
Formula Weight 379.43
Chemical Name or Material Droperidol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face, hands and any exposed skin thoroughly after handling.
EINECSNumber 208-957-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.