Learn More
Fisherbrand™ Droplet Corrugated Sample Storage Tray
Catalog No. 14127657 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
White
Catalog No. Color
14-127-657 White
1 options
Catalog No. 14-127-657
Add to Cart
Add to Cart
Affordable tray for sample organization and storage.
  • Made from corrugated polypropylene
  • Moisture resistant, significantly reducing the chance of mold
  • Stackable with other trays
  • Pack includes: 10 trays and 10 dividers
  • Trays are supplied flat-packed eliminating the hassle of stacking and reducing the amount of space needed for storage
  • Removable inserts allow outer shell to be used for storage or transport of laboratory products
  • Lightweight and sturdy to hold up to 24 full specimen containers
Specifications
Autoclavable Not Autoclavable
Color White
Holds Specimen Containers
Material Polypropylene
Positions 24
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 16 x 10.2 x 3.5 in.
Product Type BioTransport Carrier