Learn More
Sigma Aldrich Droplet Generator Chip - Four Elements Per Chip Design Fluidic 537 Pc
Catalog No. 9220131EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
92-201-31EA 1 Ea.
1 options
Catalog No. 92-201-31EA Supplier Sigma Aldrich Supplier No. 9220131EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (mul to fl) of fluids, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.Droplet generator chip - Four elements per chip design, Fluidic 537, PC is made of PC (polycarbonate). The droplet generator with design number 537 possesses a classic flow focusing geometry, perfectly suited to generate simple droplets. With four identical droplet generator units on one microfluidic chip, this device is ideal for anyone who wants to try or compare multiple experimental setups, without the need of ordering a new chip for every experiment. This chip is one of the few droplet generators devices with Luer interfaces and appropriate Luer compatible accessories are required.
Specifications
Quantity 1 Ea.
Synonym Microfluidic chip; Microparticle synthesis; Nanoparticle