Learn More
Sigma Aldrich Droplet Generator Chip - Multi Channel Design Fluidic 285 Pc
Catalog No. 9219631EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
92-196-31EA 1 Ea.
1 options
Catalog No. 92-196-31EA Supplier Sigma Aldrich Supplier No. 9219631EA
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (mul to fl) of reagents, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.Droplet generator chip - Multi channel design, Fluidic 285, PC is made of PC (polycarbonate), and has various droplet generation units. The chip features five different droplet generation units with multiple channel designs and sizes, enabling a large set of experiments. Channels/ports not in use can easily be closed by means of Mini Luer plugs. With this multichannel design several design options to generate droplets with different volumes are implemented. Main channel as well as entrance channel vary in diameter enabling a large set of experiments. Fluidic 285 features Mini Luer interfaces.
Specifications
Quantity 1 Ea.
Synonym Microfluidic chip