Learn More
Heathrow Scientific Droplet™ Phlebotomy/Sample Collection Tray Kit, Blue/Green
Description
- Multiple compartments keep a variety of supplies neatly at hand and organized
- Built in side pockets offer additional storage space
- Handle includes soft rubber grip to reduce hand and finger strain
- Each tray includes either a 13 or 16 mm OneRack™ tube rack and two disposable polystyrene inserts
- Ratcheting Handle folds out of the way for easy access, even over tall sharps containers
- Includes side pockets and supply drawer
Specifications
Specifications
| Product Type | Phlebotomy Tray |
| Material | ABS |
| Dimensions (L x W x H) | 16.8 x 10.4 x 6.7 in. (42.9 x 26.5 x 17.2 cm) |
| Height (English) | 11 in. |
| Includes | 13 mm Tube Rack Insert, 2 Disposable Inserts |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.